siRNA Details
| Gene Symbol: Rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002831.1 | NP_001002831.1 | 4, 5 | 372 | ||
| GenBank mRNA | FQ209948.1 | - | 3, 4 | 404 | ||
| FQ211619.1 | - | 4, 5 | 393 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpp21| Gene Symbol: Rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002831.1 | NP_001002831.1 | 4, 5 | 372 | ||
| GenBank mRNA | FQ209948.1 | - | 3, 4 | 404 | ||
| FQ211619.1 | - | 4, 5 | 393 | |||