siRNA Details
| Gene Symbol: Wnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002823.2 | NP_001002823.2 | 9 | 3237 | ||
| NM_001199095.1 | NP_001186024.1 | 10 | 3495 | |||
| XM_008763215.1 | XP_008761437.1 | 9 | 3303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnk1| Gene Symbol: Wnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002823.2 | NP_001002823.2 | 9 | 3237 | ||
| NM_001199095.1 | NP_001186024.1 | 10 | 3495 | |||
| XM_008763215.1 | XP_008761437.1 | 9 | 3303 | |||