siRNA Details
| Gene Symbol: Clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002807.2 | NP_001002807.1 | 3 | 397 | ||
| XM_008772745.1 | XP_008770967.1 | 3 | 398 | |||
| GenBank mRNA | BC099823.1 | - | 3 | 349 | 7932557 | |
| EU148064.1 | - | 3 | 160 | |||
| FM067949.1 | - | 3 | 397 | |||
| FM115459.1 | - | 1 | 19 | |||
| FQ219968.1 | - | 3 | 387 | |||
| FQ227136.1 | - | 3 | 183 | |||
| FQ227567.1 | - | 3 | 174 | |||
| FQ227951.1 | - | 3 | 174 | |||

Clic1
Genomic Map