siRNA Details
| Gene Symbol: Ly6g5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001934.1 | NP_001001934.1 | 2 | 172 | ||
| XM_008772747.1 | XP_008770969.1 | 3 | 859 | |||
| GenBank mRNA | HE864429.1 | - | 2 | 212 | ||
| Gene Symbol: Csnk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | HE864433.1 | - | 1829 | |||
| HE864434.1 | - | 1829 | ||||

Ly6g5b
Genomic Map