siRNA Details
Gene Symbol: G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003975.3 | NP_001003975.1 | 3 | 691 | ||
XM_006256073.2 | XP_006256135.1 | 2 | 902 | |||
GenBank mRNA | BC101859.1 | - | 3 | 671 | 7937508 |
