siRNA Details
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008823.1 | NP_001008823.1 | 5, 6 | 923 | ||
XM_006247390.2 | XP_006247452.1 | 5, 6 | 1511 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008823.1 | NP_001008823.1 | 5, 6 | 923 | ||
XM_006247390.2 | XP_006247452.1 | 5, 6 | 1511 |