siRNA Details
| Gene Symbol: St6galnac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749452.3 | XP_003749500.2 | 5 | 972 | ||
| XM_006234012.2 | XP_006234074.1 | 4 | 1134 | |||
| XM_008761787.1 | XP_008760009.1 | 3 | 948 | |||
| GenBank mRNA | AJ646871.1 | - | 3 | 703 | ||

St6galnac4
Genomic Map