siRNA Details
| Gene Symbol: Mcpt8l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573781.6 | XP_573781.1 | 2, 3 | 283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcpt8l3| Gene Symbol: Mcpt8l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573781.6 | XP_573781.1 | 2, 3 | 283 | ||