siRNA Details
| Gene Symbol: Klk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004101.1 | NP_001004101.1 | 3 | 323 | ||
| XM_008759403.1 | XP_008757625.1 | 4 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk4| Gene Symbol: Klk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004101.1 | NP_001004101.1 | 3 | 323 | ||
| XM_008759403.1 | XP_008757625.1 | 4 | 365 | |||