siRNA Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008861.2 | NP_001008861.2 | 12 | 1553 | ||
| GenBank mRNA | BC090333.1 | - | 12 | 1553 | 7190710 | |
| BC105613.1 | - | 8 | 966 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp11| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008861.2 | NP_001008861.2 | 12 | 1553 | ||
| GenBank mRNA | BC090333.1 | - | 12 | 1553 | 7190710 | |
| BC105613.1 | - | 8 | 966 | |||