siRNA Details
| Gene Symbol: Capn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002806.2 | NP_001002806.2 | 13 | 1483 | ||
| XM_006244555.2 | XP_006244617.1 | 12 | 1491 | |||
| GenBank mRNA | BC097256.1 | - | 13 | 1483 | 7457866 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn11| Gene Symbol: Capn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002806.2 | NP_001002806.2 | 13 | 1483 | ||
| XM_006244555.2 | XP_006244617.1 | 12 | 1491 | |||
| GenBank mRNA | BC097256.1 | - | 13 | 1483 | 7457866 | |