siRNA Details
| Gene Symbol: Senp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002834.1 | NP_001002834.1 | 1 | 471 | ||
| Gene Symbol: Senp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002833.1 | NP_001002833.1 | 471 | |||

Senp18
Genomic Map