siRNA Details
Gene Symbol: Spink2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008870.2 | NP_001008870.1 | 2 | 78 |
Gene Symbol: LOC100910138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751350.3 | XP_003751398.1 | 2 | 183 |
