siRNA Details
| Gene Symbol: Ece2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002815.2 | NP_001002815.2 | 1, 2 | 43 | ||
| XM_008768824.1 | XP_008767046.1 | 1, 2 | 286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ece2| Gene Symbol: Ece2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002815.2 | NP_001002815.2 | 1, 2 | 43 | ||
| XM_008768824.1 | XP_008767046.1 | 1, 2 | 286 | |||