siRNA Details
| Gene Symbol: Ece2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002815.2 | NP_001002815.2 | 17 | 1901 | ||
| XM_006248573.2 | XP_006248635.1 | 18 | 2400 | |||
| XM_008768824.1 | XP_008767046.1 | 17 | 2144 | |||
| XM_008768825.1 | XP_008767047.1 | 17 | 1888 | |||

Ece2
Genomic Map