siRNA Details
| Gene Symbol: Prss46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008865.1 | NP_001008865.1 | 2 | 157 | ||
| XM_006243993.2 | XP_006244055.1 | 3 | 397 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss46| Gene Symbol: Prss46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008865.1 | NP_001008865.1 | 2 | 157 | ||
| XM_006243993.2 | XP_006244055.1 | 3 | 397 | |||