siRNA Details
| Gene Symbol: Elavl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077651.1 | NP_001071119.1 | 2 | 314 | ||
| XM_006238591.2 | XP_006238653.1 | 2 | 475 | |||
| XM_006238592.2 | XP_006238654.1 | 2 | 790 | |||
| XM_006238593.2 | XP_006238655.1 | 2 | 223 | |||
| XM_006238594.2 | XP_006238656.1 | 2 | 499 | |||
| XM_006238595.2 | XP_006238657.1 | 2 | 474 | |||
| XM_008763893.1 | XP_008762115.1 | 2 | 401 | |||
| GenBank mRNA | BC158558.1 | - | 2 | 325 | 8371379 | |
| S83320.1 | - | 2 | 314 | |||

Elavl4
Genomic Map