siRNA Details
| Gene Symbol: Apold1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003403.1 | NP_001003403.1 | 1 | 1599 | ||
| GenBank mRNA | AY673969.1 | - | 1 | 1599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apold1| Gene Symbol: Apold1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003403.1 | NP_001003403.1 | 1 | 1599 | ||
| GenBank mRNA | AY673969.1 | - | 1 | 1599 | ||