siRNA Details
| Gene Symbol: ZW10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004724.3 | NP_004715.1 | 3 | 371 | ||
| XM_017018558.1 | XP_016874047.1 | 3 | 311 | |||
| XM_017018559.1 | XP_016874048.1 | 3 | 311 | |||
| GenBank mRNA | AK300746.1 | - | 3 | 329 | ||
| AK303871.1 | - | 2 | 50 | |||
| AK312925.1 | - | 3 | 349 | |||
| BC128264.1 | - | 3 | 299 | 40118818 | ||
| BM833775.1 | - | 3 | 371 | |||
| U54996.1 | - | 3 | 297 | |||

ZW10
Genomic Map