siRNA Details
| Gene Symbol: Thbs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013062.1 | NP_001013080.1 | 1 | 507 | ||
| GenBank mRNA | AF309630.1 | - | 1 | 507 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thbs1| Gene Symbol: Thbs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013062.1 | NP_001013080.1 | 1 | 507 | ||
| GenBank mRNA | AF309630.1 | - | 1 | 507 | ||