siRNA Details
| Gene Symbol: Repin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005893.1 | NP_001005893.1 | 1 | 133 | ||
| XM_006236423.2 | XP_006236485.1 | 2 | 1032 | |||
| XM_006236424.2 | XP_006236486.1 | 2 | 1029 | |||
| XM_006236425.2 | XP_006236487.1 | 3 | 1084 | |||
| XM_006236426.2 | XP_006236488.1 | 3 | 1082 | |||
| XM_006236427.2 | XP_006236489.1 | 3 | 1153 | |||
| XM_006236428.2 | XP_006236490.1 | 2 | 956 | |||
| XM_006236429.2 | XP_006236491.1 | 2 | 954 | |||
| GenBank mRNA | AY691175.1 | - | 1 | 133 | ||

Repin1
Genomic Map