siRNA Details
| Gene Symbol: Krt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008804.1 | NP_001008804.1 | 6 | 1302 | ||
| XM_006247391.2 | XP_006247453.1 | 6 | 7199 | |||
| XM_008768091.1 | XP_008766313.1 | 6 | 7199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt10| Gene Symbol: Krt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008804.1 | NP_001008804.1 | 6 | 1302 | ||
| XM_006247391.2 | XP_006247453.1 | 6 | 7199 | |||
| XM_008768091.1 | XP_008766313.1 | 6 | 7199 | |||