siRNA Details
| Gene Symbol: Krt32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008818.1 | NP_001008818.1 | 7 | 1319 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt32| Gene Symbol: Krt32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008818.1 | NP_001008818.1 | 7 | 1319 | ||