siRNA Details
| Gene Symbol: Krt42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008816.1 | NP_001008816.1 | 7 | 1223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt42| Gene Symbol: Krt42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008816.1 | NP_001008816.1 | 7 | 1223 | ||