siRNA Details
| Gene Symbol: Mib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005901.2 | NP_001005901.2 | 3 | 1045 | ||
| XM_006239588.1 | XP_006239650.1 | 2 | 364 | |||
| XM_006239589.2 | XP_006239651.1 | 2 | 326 | |||
| XM_008764368.1 | XP_008762590.1 | 2 | 1767 | |||
| XM_008764369.1 | XP_008762591.1 | 2 | 1767 | |||
| XM_008764370.1 | XP_008762592.1 | 2 | 1767 | |||
| XM_008764371.1 | XP_008762593.1 | 2 | 1767 | |||
| XM_008764372.1 | XP_008762594.1 | 2 | 1767 | |||

Mib2
Genomic Map