siRNA Details
| Gene Symbol: Rbm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007014.2 | NP_001007015.2 | 3 | 610 | ||
| XM_008760164.1 | XP_008758386.1 | 3 | 772 | |||
| XM_008760165.1 | XP_008758387.1 | 3 | 761 | |||
| XM_008760166.1 | XP_008758388.1 | 3 | 772 | |||
| GenBank mRNA | AY303541.1 | - | 3 | 575 | ||
| BC086416.1 | - | 3 | 610 | 7130401 | ||

Rbm4b
Genomic Map