siRNA Details
| Gene Symbol: REPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080975.1 | NP_001074444.1 | 3 | 608 | ||
| NM_004726.2 | NP_004717.2 | 3 | 608 | |||
| XM_005274625.3 | XP_005274682.1 | 3 | 572 | |||
| XM_005274626.3 | XP_005274683.1 | 3 | 572 | |||
| XM_011545603.2 | XP_011543905.1 | 3 | 572 | |||
| XM_011545604.2 | XP_011543906.1 | 3 | 572 | |||
| XM_011545605.2 | XP_011543907.1 | 3 | 572 | |||
| XM_011545606.2 | XP_011543908.1 | 3 | 572 | |||
| XM_011545609.2 | XP_011543911.1 | 3 | 220 | |||
| XM_017029955.1 | XP_016885444.1 | 3 | 572 | |||
| XM_017029956.1 | XP_016885445.1 | 3 | 572 | |||
| XM_017029957.1 | XP_016885446.1 | 3 | 572 | |||
| XM_017029958.1 | XP_016885447.1 | 3 | 572 | |||
| GenBank mRNA | AB208937.1 | - | 2 | 160 | ||
| AF010233.1 | - | 3 | 608 | |||
| AF511533.1 | - | 3 | 437 | |||
| AF512951.1 | - | 3 | 437 | |||
| AK298913.1 | - | 3 | 222 | |||

REPS2
Genomic Map