siRNA Details
| Gene Symbol: Capza2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009180.2 | NP_001009180.1 | 2, 3 | 168 | ||
| GenBank mRNA | BC101867.1 | - | 2, 3 | 168 | 7938377 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capza2| Gene Symbol: Capza2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009180.2 | NP_001009180.1 | 2, 3 | 168 | ||
| GenBank mRNA | BC101867.1 | - | 2, 3 | 168 | 7938377 | |