siRNA Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008527.1 | NP_001008527.1 | 17 | 1871 | ||
| XM_006245004.2 | XP_006245066.1 | 17 | 1899 | |||
| XM_008767121.1 | XP_008765343.1 | 17 | 1900 | |||
| XM_008767122.1 | XP_008765344.1 | 13 | 2568 | |||
| XM_008767123.1 | XP_008765345.1 | 11 | 1390 | |||
| XM_008767124.1 | XP_008765346.1 | 12 | 1566 | |||
| GenBank mRNA | AY665586.1 | - | 17 | 1871 | ||

Aox3
Genomic Map