siRNA Details
| Gene Symbol: Lmcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008562.1 | NP_001008562.1 | 2, 3 | 236 | ||
| GenBank mRNA | BC079071.1 | - | 2, 3 | 236 | 7121826 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmcd1| Gene Symbol: Lmcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008562.1 | NP_001008562.1 | 2, 3 | 236 | ||
| GenBank mRNA | BC079071.1 | - | 2, 3 | 236 | 7121826 | |