siRNA Details
| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009482.1 | NP_001009482.1 | 14 | 2011 | ||
| XM_006247204.2 | XP_006247266.1 | 14 | 2764 | |||
| XM_006247205.1 | XP_006247267.1 | 14 | 2057 | |||
| GenBank mRNA | AY353958.1 | - | 14 | 2043 | ||
| AY353959.1 | - | 14 | 2011 | |||

Mycbpap
Genomic Map