siRNA Details
| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009482.1 | NP_001009482.1 | 17, 18 | 2531 | ||
| XM_006247205.1 | XP_006247267.1 | 17, 18 | 2577 | |||
| GenBank mRNA | AY353958.1 | - | 17, 18 | 2563 | ||
| AY353959.1 | - | 17, 18 | 2531 | |||

Mycbpap
Genomic Map