siRNA Details
| Gene Symbol: Klra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009486.1 | NP_001009486.1 | 7 | 790 | ||
| XM_008763404.1 | XP_008761626.1 | 6 | 767 | |||
| GenBank mRNA | AY649835.1 | - | 7 | 790 | ||
| BC085917.1 | - | 753 | 7108565 | |||

Klra1
Genomic Map