siRNA Details
| Gene Symbol: Ly49s4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009487.1 | NP_001009487.1 | 678 | |||
| GenBank mRNA | AY649831.1 | - | 678 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly49s4| Gene Symbol: Ly49s4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009487.1 | NP_001009487.1 | 678 | |||
| GenBank mRNA | AY649831.1 | - | 678 | |||