siRNA Details
| Gene Symbol: SLC24A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254740.1 | NP_001241669.1 | 6 | 1026 | ||
| NM_001301031.1 | NP_001287960.1 | 7 | 3013 | |||
| NM_001301032.1 | NP_001287961.1 | 7 | 3049 | |||
| NM_001301033.1 | NP_001287962.1 | 10 | 3547 | |||
| NM_004727.2 | NP_004718.1 | 9 | 3264 | |||
| XM_005254778.3 | XP_005254835.1 | 10 | 3238 | |||
| XM_006720768.3 | XP_006720831.1 | 6 | 2896 | |||
| XM_011522219.1 | XP_011520521.1 | 10 | 3332 | |||
| XM_011522220.2 | XP_011520522.1 | 9 | 3184 | |||
| XM_011522221.2 | XP_011520523.1 | 9 | 3148 | |||
| XM_011522222.2 | XP_011520524.1 | 10 | 3238 | |||
| XM_017022724.1 | XP_016878213.1 | 10 | 3238 | |||
| GenBank mRNA | AB014602.1 | - | 8 | 5280 | ||
| AF026132.1 | - | 7 | 2923 | |||
| AF062921.1 | - | 8 | 2977 | |||
| AF062922.1 | - | 9 | 3264 | |||
| AK300633.1 | - | 6 | 1026 | |||
| AK304006.1 | - | 10 | 3547 | |||
| BC117263.1 | - | 7 | 2969 | 40125814 | ||
| BC143375.1 | - | 7 | 2969 | 9051880 | ||

SLC24A1
Genomic Map