siRNA Details
| Gene Symbol: Oas1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009491.1 | NP_001009491.1 | 4 | 908 | ||
| XM_008769247.1 | XP_008767469.1 | 4 | 985 | |||
| GenBank mRNA | AY196701.1 | - | 4 | 908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oas1h| Gene Symbol: Oas1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009491.1 | NP_001009491.1 | 4 | 908 | ||
| XM_008769247.1 | XP_008767469.1 | 4 | 985 | |||
| GenBank mRNA | AY196701.1 | - | 4 | 908 | ||