siRNA Details
| Gene Symbol: Oas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009493.1 | NP_001009493.1 | 826 | |||
| GenBank mRNA | AY250706.1 | - | 826 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oas3| Gene Symbol: Oas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009493.1 | NP_001009493.1 | 826 | |||
| GenBank mRNA | AY250706.1 | - | 826 | |||