siRNA Details
Gene Symbol: Ly49s7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009494.1 | NP_001009494.1 | 4 | 484 | ||
GenBank mRNA | AY649833.1 | - | 4 | 484 | ||
AY649840.1 | - | 4 | 476 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ly49s7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009494.1 | NP_001009494.1 | 4 | 484 | ||
GenBank mRNA | AY649833.1 | - | 4 | 484 | ||
AY649840.1 | - | 4 | 476 |