siRNA Details
| Gene Symbol: Ly49si1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009497.1 | NP_001009497.1 | 3 | 290 | ||
| XM_008763312.1 | XP_008761534.1 | 3 | 541 | |||
| XM_008763313.1 | XP_008761535.1 | 4 | 571 | |||
| XM_008763314.1 | XP_008761536.1 | 4 | 365 | |||
| XM_008763315.1 | XP_008761537.1 | 3 | 541 | |||
| XM_008763316.1 | XP_008761538.1 | 3 | 541 | |||
| XM_008763317.1 | XP_008761539.1 | 2 | 451 | |||
| XM_008763318.1 | XP_008761540.1 | 2 | 451 | |||
| XM_008763319.1 | XP_008761541.1 | 3 | 541 | |||
| GenBank mRNA | AY659935.1 | - | 3 | 290 | ||

Ly49si1
Genomic Map