siRNA Details
| Gene Symbol: Eogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009502.1 | NP_001009502.1 | 15 | 1681 | ||
| XM_008763144.1 | XP_008761366.1 | 12 | 1355 | |||
| XM_008763145.1 | XP_008761367.1 | 13 | 1267 | |||
| XM_008763146.1 | XP_008761368.1 | 11 | 1147 | |||
| XM_008763147.1 | XP_008761369.1 | 12 | 1412 | |||
| GenBank mRNA | AJ868236.1 | - | 15 | 1681 | ||

Eogt
Genomic Map