siRNA Details
Gene Symbol: Pfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009503.1 | NP_001009503.1 | 2 | 1125 | ||
GenBank mRNA | AY682392.1 | - | 2 | 1125 | ||
BC107465.1 | - | 2 | 1257 | 7453892 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009503.1 | NP_001009503.1 | 2 | 1125 | ||
GenBank mRNA | AY682392.1 | - | 2 | 1125 | ||
BC107465.1 | - | 2 | 1257 | 7453892 |