siRNA Details
| Gene Symbol: Fbxl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009504.1 | NP_001009504.1 | 1 | 553 | ||
| XM_006246020.2 | XP_006246082.1 | 2 | 871 | |||
| GenBank mRNA | AY823669.1 | - | 1 | 553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl16| Gene Symbol: Fbxl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009504.1 | NP_001009504.1 | 1 | 553 | ||
| XM_006246020.2 | XP_006246082.1 | 2 | 871 | |||
| GenBank mRNA | AY823669.1 | - | 1 | 553 | ||