siRNA Details
| Gene Symbol: Vom1r111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009522.1 | NP_001009522.1 | 1 | 464 | ||
| GenBank mRNA | AY510366.1 | - | 1 | 464 | ||
| Gene Symbol: Vom1r17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167572.1 | NP_001161044.1 | 464 | |||

Vom1r111
Genomic Map