siRNA Details
| Gene Symbol: Vom1r86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009527.1 | NP_001009527.1 | 1 | 757 | ||
| GenBank mRNA | AY510308.1 | - | 1 | 757 | ||
| Gene Symbol: Vom1r87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009529.1 | NP_001009529.1 | 1 | 757 | ||
| GenBank mRNA | AY510311.1 | - | 1 | 757 | ||

Vom1r86
Genomic Map