siRNA Details
| Gene Symbol: Vom1r84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009528.1 | NP_001009528.1 | 1 | 240 | ||
| XM_008762962.1 | XP_008761184.1 | 2 | 313 | |||
| GenBank mRNA | AY510312.1 | - | 1 | 240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r84| Gene Symbol: Vom1r84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009528.1 | NP_001009528.1 | 1 | 240 | ||
| XM_008762962.1 | XP_008761184.1 | 2 | 313 | |||
| GenBank mRNA | AY510312.1 | - | 1 | 240 | ||