siRNA Details
Gene Symbol: Vom1r87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009529.1 | NP_001009529.1 | 1 | 102 | ||
GenBank mRNA | AY510311.1 | - | 1 | 102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vom1r87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009529.1 | NP_001009529.1 | 1 | 102 | ||
GenBank mRNA | AY510311.1 | - | 1 | 102 |