siRNA Details
| Gene Symbol: Vom1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008903.1 | NP_001008903.1 | 1 | 613 | ||
| GenBank mRNA | AY510349.1 | - | 1 | 613 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r2| Gene Symbol: Vom1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008903.1 | NP_001008903.1 | 1 | 613 | ||
| GenBank mRNA | AY510349.1 | - | 1 | 613 | ||