siRNA Details
| Gene Symbol: Vom1r40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008904.1 | NP_001008904.1 | 1 | 150 | ||
| GenBank mRNA | AY510277.1 | - | 1 | 150 | ||
| Gene Symbol: LOC102551504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228159.1 | XP_006228221.1 | 1 | 150 | ||

Vom1r40
Genomic Map