siRNA Details
| Gene Symbol: Vom1r41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008909.1 | NP_001008909.1 | 1 | 80 | ||
| GenBank mRNA | AY510299.1 | - | 1 | 80 | ||
| Gene Symbol: Vom1r107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008908.1 | NP_001008908.1 | 1 | 80 | ||
| GenBank mRNA | AY510300.1 | - | 1 | 80 | ||

Vom1r41
Genomic Map