siRNA Details
| Gene Symbol: Vom1r74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008911.1 | NP_001008911.1 | 1 | 440 | ||
| GenBank mRNA | AY510285.1 | - | 1 | 440 | ||
| Gene Symbol: LOC100909399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749767.1 | XP_003749815.1 | 1 | 440 | ||
| Gene Symbol: Vom1r73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008922.1 | NP_001008922.1 | 1 | 440 | ||
| GenBank mRNA | AY510289.1 | - | 1 | 440 | ||
| Gene Symbol: Vom1r78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008924.1 | NP_001008924.1 | 1 | 440 | ||
| GenBank mRNA | AY510292.1 | - | 1 | 440 | ||
| Gene Symbol: Vom1r79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009508.1 | NP_001009508.1 | 1 | 383 | ||
| GenBank mRNA | AY510284.1 | - | 1 | 383 | ||

Vom1r74
Genomic Map